Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMNB1, Mouse, Clone: ZL-5, Abnova™

Catalog No. 89110610 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-610 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-610 Supplier Abnova Corporation Supplier No. MAB1761

Mouse monoclonal antibody raised against recombinant LMNB1.

The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. This gene encodes one of the two B type proteins, B1. [provided by RefSeq

Specifications

Antigen LMNB1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone ZL-5
Conjugate Unconjugated
Description lamin B1
Formulation In PBS (50% glycerol, 1mg/mL BSA, 1.5mM sodium azide)
Gene LMNB1
Gene Alias ADLD, LMN, LMN2, LMNB, MGC111419
Gene Symbols LMNB1
Host Species Mouse
Immunogen Recombinant protein corresponding to human LMNB1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4001
Test Specificity This antibody recognizes the 68kDa lamin B1 isoform.
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.