Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO2 (M05J), Mouse anti-Human, Clone: 4D3, Abnova™

Catalog No. 89104849 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-849 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-849 Supplier Abnova Corporation Supplier No. H00004005M05J
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant LMO2.

LMO2 encodes a cysteine-rich, two LIM-domain protein that is required for yolk sac erythropoiesis. The LMO2 protein has a central and crucial role in hematopoietic development and is highly conserved. The LMO2 transcription start site is located approximately 25kb downstream from the 11p13 T-cell translocation cluster (11p13 ttc), where a number T-cell acute lymphoblastic leukemia-specific translocations occur. Alternative splicing results in multiple transcript variants encoding different isoforms

Sequence: PVDEVLQIPPSLLTCGGCQQNIGDRYFLKAIDQYWHEDCLSCDLCGCRLGEVGRRLYYKLGRKLCRRDYLRLFGQDGLCASCDKRIR

Specifications

Antigen LMO2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant LMO2.
Formulation PBS with no preservative; pH 7.4
Gene LMO2
Gene Accession No. NM_005574
Gene Alias RBTN2/RBTNL1/RHOM2/TTG2
Gene Symbols LMO2
Host Species Mouse
Immunogen LMO2 (NP_005565,16 a.a. ∽ 102 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4005
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.