Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMTK3, Mouse, Clone: 2H6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LMTK3.
Supplier: Abnova Corporation H00114783M02A
Description
Sequence: VLVNGGLTPPKSEDKVSENGGLRFPRNTERPPETGPWRAPGPWEKTPESWGPAPTIGEPAPETSLERAPAPSAVVSSRNGGETAPGPLGPAPKNGTLEPGSpecifications
LMTK3 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
XM_055866 | |
LMTK3 | |
LMTK3 (XP_055866,1151 a.a. ∽ 1250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
114783 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
2H6 | |
Mouse monoclonal antibody raised against a partial recombinant LMTK3. | |
LMTK3 | |
KIAA1883/LMR3/TYKLM3 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction