Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP1, Mouse, Clone: A2Mr alpha-2, Abnova™

Catalog No. 89112162 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-162 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-112-162 Supplier Abnova Corporation Supplier No. MAB5197
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against LRP1.

Specifications

Antigen LRP1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A2Mr alpha-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against LRP1.
Dilution Flow Cytometry (10ul of the suggested working dilution (1:50-1:100) to label 106 cells or 100ul ) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LRP1
Gene Alias A2MR/APOER/APR/CD91/FLJ16451/IGFBP3R/LRP/MGC88725/TGFBR5
Gene Symbols LRP1
Host Species Mouse
Immunogen Purified LRP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4035
Test Specificity This antibody recognizes human CD91, a type I membrane protein which is also known as the alpha 2 macroglobulin receptor (alpha 2MR). It recognizes an epitope in the extracellular region of the 500kD alpha chain.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.