Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP1, Mouse, Clone: MA8G1, Abnova™

Catalog No. 89111614 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-614 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-614 Supplier Abnova Corporation Supplier No. MAB3822
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against LRP1.

Specifications

Antigen LRP1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MA8G1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against LRP1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium phosphate buffer, 150mM NaCl, pH 7.4
Gene LRP1
Gene Alias A2MR/APOER/APR/CD91/FLJ16451/IGFBP3R/LRP/MGC88725/TGFBR5
Gene Symbols LRP1
Host Species Mouse
Immunogen Human 515 kDa low density LRP1.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4035
Test Specificity This monoclonal antibody is produced in a mouse ascites and is specific to the human heavy chain 515 KDa low density lipoprotein receptor-related protein (LRP). It is a useful marker for the characterization of a subset of myelo-monocytic subtypes of chronic and acute leukemia (CD91).
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.