Learn More
LRP1B, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant LRP1B.
Supplier: Abnova Corporation H00053353A01
Description
LRP1B belongs to the low density lipoprotein (LDL) receptor gene family. These receptors play a wide variety of roles in normal cell function and development due to their interactions with multiple ligands (Liu et al., 2001 [PubMed 11384978]).[supplied by OMIM
Sequence: VHCQELLSNCQQLNCQYKCTMVRNSTRCYCEDGFEITEDGRSCKDQDECAVYGTCSQTCRNTHGSYTCSCVEGYLMQPDNRSCKAKIEPTSpecifications
LRP1B | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant LRP1B. | |
LRP1B | |
LRP-DIT/LRPDIT | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_018557 | |
LRP1B | |
LRP1B (NP_061027, 111 a.a. ∽ 200 a.a) partial recombinant protein with GST tag. | |
RUO | |
53353 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only