Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP5, Mouse, Clone: 1E9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LRP5.
Supplier: Abnova Corporation H00004041M01A
Description
Sequence: RDVRLVDAGGVKLESTIVVSGLEDAAAVDFQFSKGAVYWTDVSEEAIKQTYLNQTGAAVQNVVISGLVSPDGLACDWVGKKLYWTDSETNSpecifications
LRP5 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_002335 | |
LRP5 | |
LRP5 (NP_002326.1,41 a.a. ∽ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
4041 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
1E9 | |
Mouse monoclonal antibody raised against a partial recombinant LRP5. | |
LRP5 | |
BMND1/EVR1/EVR4/HBM/LR3/LRP7/OPPG/OPS/OPTA1/VBCH2 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction