Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRPAP1, Mouse, Clone: MA7F1, Abnova™

Catalog No. 89111611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-611 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-611 Supplier Abnova Corporation Supplier No. MAB3819
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against LRPAP1.

Specifications

Antigen LRPAP1
Applications Western Blot
Classification Monoclonal
Clone MA7F1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against LRPAP1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM Tris-HCl, 100mM NaCl, pH 7.4
Gene LRPAP1
Gene Alias A2MRAP/A2RAP/HBP44/MGC138272/MRAP/RAP
Gene Symbols LRPAP1
Host Species Mouse
Immunogen Human LRPAP1.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4043
Test Specificity This is a mouse monoclonal antibody is specific to the 38 KDa low density lipoprotein receptor-associated protein (RAP). It does not cross react with mouse or rabbit and is suitable for Western blot.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.