Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM16, Mouse, Clone: 4E3-3A4, Abnova™

Catalog No. 89107362 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-362 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-362 Supplier Abnova Corporation Supplier No. H00080153M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant LSM16.

EDC3 is associated with an mRNA-decapping complex required for removal of the 5-prime cap from mRNA prior to its degradation from the 5-prime end (Fenger-Gron et al., 2005 [PubMed 16364915]).[supplied by OMIM

Sequence: MATDWLGSIVSINCGDSLGVYQGRVSAVDQVSQTISLTRPFHNGVKCLVPEVTFRAGDITELKILEIPGPGDNQHFGDLHQTELGPSGAGCQVGINQNGTGKFVKKPASSSSAPQNIPKRTDVKSQDVAVSPQQQQCSKSYVDRHMESLSQSKSFRRRHNSWSSSSRHPNQATPKKSGLKNGQMKNKDDECFGDDIEEIPDTDFDFEGNLALFDKAAVFEEIDTYERRSGTRSRGIPNERPTRYRHDENILESEPIVYRRIIVPHNVSKEFCTDSGLVVPSISYELHKKLLSVAEKHGLTLERRLEMTGVCASQMALTLLGGPNRLNPKNVHQRPTVALLCGPHVKGAQGISCGRHLANHDVQVILFLPNFVKMLESITNELSLFSKTQGQQVSSLKDLPTSPVDLVINCLDCPENVFLRDQPWYKAAVAWANQNRAPVLSIDPPVHEVEQGIDAKWSLALGLPLPLGEHAGRIYLCDIGIPQQVFQEVGINYHSPFGCKFVIPLHSA

Specifications

Antigen LSM16
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E3-3A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant LSM16.
Formulation ascites with no preservative
Gene EDC3
Gene Accession No. BC011534
Gene Alias FLJ21128/FLJ31777/LSM16/YJDC/YJEFN2
Gene Symbols EDC3
Host Species Mouse
Immunogen LSM16 (AAH11534,1 a.a. ∽ 508 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80153
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.