Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM2, Mouse, Clone: AT2B2, Abnova™

Catalog No. 89109360 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-360 Supplier Abnova Corporation Supplier No. MAB11192
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant LSM2.

Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM

Specifications

Antigen LSM2
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT2B2
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LSM2.
Dilution Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline (pH 7.4) with 0.09% sodium azide.
Gene LSM2
Gene Alias C6orf28/G7b/YBL026W/snRNP
Gene Symbols LSM2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-95 of human LSM2.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57819
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.