Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LST1, Mouse, Clone: LST1/02, Abnova™

Catalog No. 89112892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-892 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-892 Supplier Abnova Corporation Supplier No. MAB6480
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of LST1.

Sequence: SSEGPDLRGRDKRGT

Specifications

Antigen LST1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LST1/02
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of LST1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LST1
Gene Alias B144/D6S49E/LST-1/MGC119006/MGC119007
Gene Symbols LST1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human LST1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7940
Test Specificity This antibody reacts with an extracellular epitope of LST1, an approximately 6-11 KDa protein expressed as various transmembrane or soluble forms. It migrates on SDS PAGE gels asapproximately 25-28 KDa molecule.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.