Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTF, Mouse, FITC, Clone: NI 25, Abnova™

Catalog No. 89108386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-386 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-386 Supplier Abnova Corporation Supplier No. MAB10069
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against LTF.

This gene is a member of the transferrin family of genes and its protein product is found in the secondary granules of neutrophils. The protein is a major iron-binding protein in milk and body secretions with an antimicrobial activity, making it an important component of the nonspecific immune system. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. [provided by RefSeq

Specifications

Antigen LTF
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone NI 25
Conjugate FITC
Description Mouse monoclonal antibody raised against LTF.
Dilution Immunohistochemistry (1:10) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.2
Gene LTF
Gene Alias GIG12/HLF2/LF
Gene Symbols LTF
Host Species Mouse
Immunogen Purified LTF from human milk.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4057
Test Specificity The antiserum does not react with any other component of the human milk or plasma protein as tested. This antiserum has not been tested for cross-reactivity with other species.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.5mL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.