Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luciferase, Mouse, Clone: Luci 21 1-107, Abnova™

Catalog No. 89135165 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-165 Supplier Abnova Corporation Supplier No. MAB2322
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native luciferase.

Specifications

Antigen Luciferase
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone Luci 21 1-107
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native luciferase.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Host Species Mouse
Immunogen Photinus pyralis., Native purified luciferase protein from firefly
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes a band at ∼61 KDa on Western Blot. specific to luciferase, recognizing a peptide consisting of the first 258 amino acids. Further epitope mapping has not been done at this time.
Target Species Firefly
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.