Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lung carcinoma Cluster 1, Mouse, Clone: MOC-1, Abnova™
Mouse monoclonal antibody raised against native Lung carcinoma Cluster 1.
Supplier: Abnova Corporation MAB3546
Specifications
Lung carcinoma Cluster 1 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
3 mL | |
Primary | |
Bovine, Human | |
Liquid |
Immunohistochemistry | |
MOC-1 | |
Mouse monoclonal antibody raised against native Lung carcinoma Cluster 1. | |
In phosphate buffer, pH 7.3 (0.09% sodium azide) | |
Native purified from SCLC-derived cell line. | |
RUO | |
This antibody reacts with cluster 1 membrane protein 125-145 KDa. | |
Store at 4°C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction