Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Luteinizing hormone, Mouse, Clone: 1/2D7 1D9, Abnova™

Catalog No. 89135504 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-504 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-504 Supplier Abnova Corporation Supplier No. MAB3688
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Luteinizing hormone (LH).

Specifications

Antigen Luteinizing hormone
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1/2D7 1D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Luteinizing hormone (LH).
Dilution The optimal working dilution should be determined by the end user.
Formulation In BBS
Host Species Mouse
Immunogen Luteinizing hormone
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibosy recognizes natural and recombinant human Luteinizing Hormone.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.