Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Luteinizing hormone, Mouse, Clone: 2/22E2 2H4, Abnova™
Mouse monoclonal antibody raised against recombinant Luteinizing hormone (LH).
Supplier: Abnova Corporation MAB3689
Specifications
Luteinizing hormone | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Human | |
Liquid |
ELISA, Radioimmune Assays (RIA) | |
2/22E2 2H4 | |
Mouse monoclonal antibody raised against recombinant Luteinizing hormone (LH). | |
In BBS | |
Luteinizing hormone | |
RUO | |
This antibosy recognizes natural and recombinant human Luteinizing Hormone. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction