Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY9, Mouse, Clone: HLy9.25, Abnova™

Catalog No. 89113524 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-524 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-524 Supplier Abnova Corporation Supplier No. MAB8703
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant LY9.

LY9 belongs to the SLAM family of immunomodulatory receptors (see SLAMF1; MIM 603492) and interacts with the adaptor molecule SAP (SH2D1A; MIM 300490) (Graham et al., 2006 [PubMed 16365421]).[supplied by OMIM

Specifications

Antigen LY9
Applications Flow Cytometry, Functional Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone HLy9.25
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant LY9.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LY9
Gene Alias CD229/SLAMF3/hly9/mLY9
Gene Symbols LY9
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human LY9.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4063
Test Specificity This antibody is specific to LY9.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.