Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY96, Mouse, Clone: 1A2E3, Abnova™

Catalog No. 89111125 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-125 Supplier Abnova Corporation Supplier No. MAB2753
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant LY96.

The MD-2 protein appears to associate with toll-like receptor 4 on the cell surface and confers responsiveness to lipopolysaccyaride (LPS), thus providing a link between the receptor and LPS signaling. [provided by RefSeq

Specifications

Antigen LY96
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1A2E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant LY96.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene LY96
Gene Alias MD-2/MD2
Gene Symbols LY96
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 2-160 of human LY96.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23643
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.