Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYN, Mouse, Clone: 2H8D7, Abnova™

Catalog No. 89112282 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-282 Supplier Abnova Corporation Supplier No. MAB5531
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant LYN.

Specifications

Antigen LYN
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H8D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LYN.
Dilution ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene LYN
Gene Alias FLJ26625/JTK8
Gene Symbols LYN
Host Species Mouse
Immunogen Recombinant protein corresponding to human LYN.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4067
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.