Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYN, Mouse, Clone: LYN-01, Abnova™

Catalog No. 89108277 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-277 Supplier Abnova Corporation Supplier No. MAB0949
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant LYN.

Specifications

Antigen LYN
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LYN-01
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LYN.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LYN
Gene Alias FLJ26625/JTK8
Gene Symbols LYN
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 8-238 of human LYN.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4067
Test Specificity This antibody reacts with Lyn (p56/p53), a non-receptor Src-family tyrosine kinase expressed in hematopoietic tissues.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.