Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LYVE1, Mouse, Clone: 4G1, Abnova™

Catalog No. 89108869 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-869 Supplier Abnova Corporation Supplier No. MAB1067
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant LYVE1.

This gene encodes a type I integral membrane glycoprotein. The encoded protein acts as a receptor and binds to both soluble and immobilized hyaluronan. This protein may function in lymphatic hyaluronan transport and have a role in tumor metastasis. [provided by RefSeq

Specifications

Antigen LYVE1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LYVE1.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LYVE1
Gene Accession No. NM_006691
Gene Alias CRSBP-1/HAR/LYVE-1/XLKD1
Gene Symbols LYVE1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 25-235 of human LYVE1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10894
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.