Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human M-CSF R/CD115 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
$241.00 - $2020.00
Specifications
Antigen | M-CSFR/CD115 |
---|---|
Clone | 61701 |
Dilution | Western Blot 2 ug/mL, Neutralization 4-16 ng/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3291SP
![]() |
R&D Systems
MAB3291SP |
25 μg |
Each for $241.00
|
|
|||||
MAB3291100
![]() |
R&D Systems
MAB3291100 |
100 μg |
Each for $573.00
|
|
|||||
MAB3291500
![]() |
R&D Systems
MAB3291500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for Western Blot, Neutralization.Specifications
M-CSFR/CD115 | |
Western Blot 2 ug/mL, Neutralization 4-16 ng/mL | |
Monoclonal | |
Mouse | |
Human | |
P07333.2 | |
1436 | |
Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115 Ile20-Glu512 (Pro54Ala) Accession # P07333.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human M-CSF R/CD115 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) GM-CSF R alpha or rhGM-CSF R beta is observed. |
61701 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
CSF1R | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title