Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M13/fd/F1 Filamentous Phages, g8p, Mouse, Biotin, Clone: R7129.7, Abnova™

Catalog No. 89135971 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-971 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-971 Supplier Abnova Corporation Supplier No. MAB5439
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against M13/fd/F1 Filamentous Phages, g8p.

Specifications

Antigen M13/fd/F1 Filamentous Phages, g8p
Applications ELISA, Western Blot
Classification Monoclonal
Clone R7129.7
Conjugate Biotin
Description Mouse monoclonal antibody raised against M13/fd/F1 Filamentous Phages, g8p.
Dilution ELISA (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.5% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen fd phages from Escherichia coli (F+ strain (JM109)).
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Binds to an epitope on pVIII (phage coat protein) covering the N-terminal region of g8p AEGDDPAKAAFDSLQASAT.
Target Species E. coli, Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.