Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage marker, Mouse, Clone: 3A5, Abnova™

Catalog No. 89135509 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-509 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-509 Supplier Abnova Corporation Supplier No. MAB3756
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Macrophage marker.

Specifications

Antigen Macrophage marker
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 3A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Macrophage marker.
Dilution Immunohistochemistry (1:20) The optimal working dilution should be determined by the end user.
Formulation In serum-free culture supernatant (1% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Native purified from fragments of human spleen depleted of lymphocytes and erythrocytes.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts specifically with macrophages, monophages, and histiocytic cells in normal and diseased tissue. Dendritic cells (interdigitating cells, Langerhans' cells, and microglia), myeloid, lymphoid, and epithelial cells not stained with 3A5.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.