Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Macrophage specific antigen, Mouse, Clone: F-6-J, Abnova™

Catalog No. 89135905 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-905 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-905 Supplier Abnova Corporation Supplier No. MAB5180

Mouse monoclonal antibody raised against native Macrophage specific antigen.

Specifications

Antigen Macrophage specific antigen
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone F-6-J
Conjugate Unconjugated
Formulation In PBS (50% glycerol, 0.01% sodium azide)
Host Species Mouse
Immunogen Native purified from WAG/Rij lymphnode cells.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The monoclonal antibody recognizes rat macrophages and is reactive with all common inbred strains. F-6-J is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from Balb/c mice immunized with WAG/Rij lymphnode cells.
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.