Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD1L1, Mouse, Clone: 9B10, Abnova™

Catalog No. 89110714 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-714 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-714 Supplier Abnova Corporation Supplier No. MAB1930
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant.

MAD1L1 is a component of the mitotic spindle-assembly checkpoint that prevents the onset of anaphase until all chromosome are properly aligned at the metaphase plate. MAD1L1 functions as a homodimer and interacts with MAD2L1. MAD1L1 may play a role in cell cycle control and tumor suppression. Three transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen MAD1L1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant.
Dilution Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene MAD1L1
Gene Alias HsMAD1/MAD1/PIG9/TP53I9/TXBP181
Gene Symbols MAD1L1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human MAD1L1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8379
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.