Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAD2L1, Mouse, Clone: KD6.17D10.2G9, Abnova™

Catalog No. 89113662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-662 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-662 Supplier Abnova Corporation Supplier No. MAB8965
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MAD2L1.

MAD2L1 is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. MAD2L1 is related to the MAD2L2 gene located on chromosome 1. A MAD2 pseudogene has been mapped to chromosome 14. [provided by RefSeq

Specifications

Antigen MAD2L1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KD6.17D10.2G9
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MAD2L1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene MAD2L1
Gene Alias HSMAD2/MAD2
Gene Symbols MAD2L1
Host Species Mouse
Immunogen Recombinant protein corresponding to human MAD2L1.
Purification Method Purified ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4085
Test Specificity Western blot with a variety of human cell lines confirms that the antibody is specific for human Mad2; other species specificity has not yet been determined.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.