Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA11, Mouse, Clone: 1C11, Abnova™

Catalog No. 89104879 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-879 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-879 Supplier Abnova Corporation Supplier No. H00004110M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MAGEA11.

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MPLEQRSQHCKPEEGLQAQEEDLGLVGAQALQAEEQEAAFFSSTLNVGTLEELPAAESPSPPQSPQEESFSPTAMDAIFGSLSDEGSGSQEKEGPSTSPDLIDPESFSQDILHDKIIDLVHLLLRKYRVKGLITKAEMLGSVIKNYEDYFPEIFREASVCMQLLFGIDVKEVDPTSHSYVLVTSLNLSYDGIQCNEQSMPKSGLLIIVLGVIFMEGNCIPEEVMWEVLSIMGVYAGREHFLFGEPKRLLTQNWVQEKYLVYRQVPGTDPACYEFLWGPRAHAETSKMKVLEYIANANGRDPTSYPSLYEDALREEGEGV

Specifications

Antigen MAGEA11
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant MAGEA11.
Formulation ascites with no preservative
Gene MAGEA11
Gene Accession No. BC004479.1
Gene Alias MAGE-11/MAGE11/MAGEA-11/MGC10511
Gene Symbols MAGEA11
Host Species Mouse
Immunogen MAGEA11 (AAH04479.1,1 a.a. ∽ 319 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4110
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.