Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAML1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106154 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-154 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-154 Supplier Abnova Corporation Supplier No. H00009794A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MAML1.

This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway. [provided by RefSeq

Sequence: AEQEKQQFQRHLTRPPPQYQDPTQGSFPQQVGQFTGSSAAVPGMNTLGPSNSSCPRVFPQAGNLMPMGPGHASVSSLPTNSGQQDRGVAQFPGSQNMPQS

Specifications

Antigen MAML1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MAML1.
Formulation 50% glycerol
Gene MAML1
Gene Accession No. NM_014757
Gene Alias KIAA0200/Mam-1/Mam1
Gene Symbols MAML1
Host Species Mouse
Immunogen MAML1 (NP_055572, 655 a.a. ∽ 754 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9794
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.