Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1B, Mouse, Clone: 3G5, Abnova™

Catalog No. 89111578 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-578 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-578 Supplier Abnova Corporation Supplier No. MAB3746

Mouse monoclonal antibody raised against native MAP1B.

Specifications

Antigen MAP1B
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native MAP1B.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene MAP1B
Gene Symbols MAP1B
Host Species Mouse
Immunogen Native purified MAP1B from bovine brain.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 514739
Test Specificity Clone 3G5 recognizes a protein of 320 KDa identified as MAP1B, MAP5, MAP1.2, MAP1(x) or MAP1X. This antibody reacts with MAP1B. Highly specific to MAP1B and shows no cross-reactivity with other MAPs, tau and tubulin. Does not react with chicken.
Target Species Bovine, Human, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.