Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89113562 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-562 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-562 Supplier Abnova Corporation Supplier No. MAB8746
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MAP2.

This gene encodes a protein that belongs to the microtubule-associated protein family. The proteins of this family are thought to be involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dentrites, implicating a role in determining and stabilizing dentritic shape during neuron development. A number of alternatively spliced variants encoding distinct isforms have been described. [provided by RefSeq

Specifications

Antigen MAP2
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MAP2.
Dilution Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide, 1% BSA, 50% glycerol)
Gene MAP2
Gene Accession No. NM_031846.2
Gene Alias DKFZp686I2148/MAP2A/MAP2B/MAP2C
Gene Symbols MAP2
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus residues of human MAP2.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4133
Test Specificity This antibody is specific to MAP2.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.