Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2K4, Mouse, Clone: 2D10D8, 4G11B2, Abnova™

Catalog No. 89112283 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-283 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-283 Supplier Abnova Corporation Supplier No. MAB5532
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MAP2K4.

This gene encodes a dual specificity protein kinase that belongs to the Ser/Thr protein kinase family. This kinase is a direct activator of MAP kinases in response to various environmental stresses or mitogenic stimuli. It has been shown to activate MAPK8/JNK1, MAPK9/JNK2, and MAPK14/p38, but not MAPK1/ERK2 or MAPK3/ERK3. This kinase is phosphorylated, and thus activated by MAP3K1/MEKK. The knockout studies in mice suggested the roles of this kinase in mediating survival signal in T cell development, as well as in the organogenesis of liver. [provided by RefSeq

Specifications

Antigen MAP2K4
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2D10D8, 4G11B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MAP2K4.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene MAP2K4
Gene Alias JNKK/JNKK1/MAPKK4/MEK4/MKK4/PRKMK4/SEK1/SERK1
Gene Symbols MAP2K4
Host Species Mouse
Immunogen Recombinant protein corresponding to human MAP2K4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6416
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.