Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-MAP2K6, Clone: 3H12C8,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS810
Description
Specifically detects MAP2K6 clone: 3H12C8 in Human, Mouse samples, and it is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blotting
Dual specificity mitogen-activated protein kinase kinase 6 (MP2K6) is a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. MP2K6 phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
Specifications
MAP2K6 | |
Monoclonal | |
Mouse monoclonal IgG1 ascitic fluid containing 0.03% sodium azide. | |
MAP2K6; MEK6; MKK6; PRKMK6; SKK3 | |
Purified recombinant fragment of human MAP2K6 expressed in E. coli. | |
100 μL | |
Signaling | |
Human, Mouse | |
Ascites |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
3H12C8 | |
P52564 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction