Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP3K11, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104931 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-931 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-931 Supplier Abnova Corporation Supplier No. H00004296A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MAP3K11.

The protein encoded by this gene is a member of the serine/threonine kinase family. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42. [provided by RefSeq

Sequence: PPPGTSRSAPGTPGTPRSPPLGLISRPRPSPLRSRIDPWSFVSAGPRPSPLPSPQPAPRRAPWTLFPDSDPFWDSPPANPFQGGPQDCRAQTKDMGAQAPWVPEAGP

Specifications

Antigen MAP3K11
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MAP3K11.
Formulation 50% glycerol
Gene MAP3K11
Gene Accession No. NM_002419
Gene Alias MGC17114/MLK-3/MLK3/PTK1/SPRK
Gene Symbols MAP3K11
Host Species Mouse
Immunogen MAP3K11 (NP_002410, 741 a.a. ∽ 847 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4296
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.