Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-MAP3K2, Clone: 4B4,

Catalog No. MABS816MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS816MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS816MI Supplier MilliporeSigma™ Supplier No. MABS816
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects MAP3K2 clone: 4B4 in Human samples, and it is validated for ELISA, Immunocytochemistry, Western Blotting

Mitogen-activated protein kinase kinase kinase 2 (MAP3K2) is a member of serine/threonine protein kinase family. MAP3K2 preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. MAP3K2 has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP3K2
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4B4
Formulation Mouse monoclonal IgG1 ascitic fluid containing 0.03% sodium azide.
Gene Accession No. Q9Y2U5
Gene Symbols MAP3K2; MAPKKK2; MEKK2
Host Species Mouse
Immunogen Purified recombinant fragment of human ERK3 expressed in E. coli
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.