Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK1, Mouse, Clone: 1D1, Abnova™

Catalog No. 89105203 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-203 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-203 Supplier Abnova Corporation Supplier No. H00005594M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MAPK1.

The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets. Two alternatively spliced transcript variants encoding the same protein, but differing in the UTRs, have been reported for this gene. [provided by RefSeq

Sequence: RNYLLSLPHKNKVPWNRLFPNADSKALDLLDKMLTFNPHKRIEVEQALAHPYLEQYYDPSDEPIAEAPFKFDMELDDLPKEKLKELIFEETARFQPGYRS

Specifications

Antigen MAPK1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MAPK1.
Formulation ascites with no preservative
Gene MAPK1
Gene Accession No. BC017832
Gene Alias ERK/ERK2/ERT1/MAPK2/P42MAPK/PRKM1/PRKM2/p38/p40/p41/p41mapk
Gene Symbols MAPK1
Host Species Mouse
Immunogen MAPK1 (AAH17832,261 a.a. ∽ 360 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5594
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.