Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK1/MAPK3, Mouse, Clone: SB46b, Abnova™

Catalog No. 89111357 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-357 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-357 Supplier Abnova Corporation Supplier No. MAB3337
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MAPK1/MAPK3.

Specifications

Antigen MAPK1/MAPK3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SB46b
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MAPK1/MAPK3.
Dilution Western Blot (1-2 ug/mL) Immunoprecipitation (10-15ug) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.0
Gene Alias ERK1/HS44KDAP/HUMKER1A/MGC20180/P44ERK1/P44MAPK/PRKM3
Gene Symbols MAPK3
Host Species Mouse
Immunogen Recombinant protein corresponding to human MAPK1/MAPK3.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5595/5594
Test Specificity human Erk 1 and human Erk2.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.