Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK14, Mouse, Clone: 9F12, Abnova™

Catalog No. 89136258 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-258 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-258 Supplier Abnova Corporation Supplier No. MAB6637
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant MAPK14.

Specifications

Antigen MAPK14
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant MAPK14.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Host Species Mouse
Immunogen Recombinant protein corresponding to full length MAPK14.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects a ∼38KDa protein corresponding to the molecular mass of p38alpha MAPK on SDS PAGE immunoblots.
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.