Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mapk14, Mouse, Clone: M138, Abnova™

Catalog No. 89110400 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-400 Supplier Abnova Corporation Supplier No. MAB1384
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Mapk14.

Specifications

Antigen Mapk14
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone M138
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Mapk14.
Dilution ELISA (1:2000) Immunocytochemistry (1:500) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide)
Gene Mapk14
Gene Alias CSBP/CSPB1/Csbp1/Csbp2/Exip/Hog/MGC105413/Mxi2/Prkm14/Prkm15/RK/Sapk2A/p38/p38Hog/p38alpha
Gene Symbols Mapk14
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of rat Mapk14.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81649
Test Specificity This sequence is identical in human and mouse p38a.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.