Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105204 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-204 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-204 Supplier Abnova Corporation Supplier No. H00005596A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MAPK4.

Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets. [provided by RefSeq

Sequence: ACRKVAVKKIALSDARSMKHALREIKIIRRLDHDNIVKVYEVLGPKGTDLQGELFKFSVAYIVQEYMETDLARLLEQGTLAEEHAKLFMYQLLRGLKYIHSANVLHRDLK

Specifications

Antigen MAPK4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MAPK4.
Formulation 50% glycerol
Gene MAPK4
Gene Accession No. NM_002747
Gene Alias ERK3/Erk4/PRKM4/p63MAPK
Gene Symbols MAPK4
Host Species Mouse
Immunogen MAPK4 (NP_002738, 42 a.a. ∽ 151 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5596
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.