Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPT, Mouse, Clone: TAU-5, Abnova™

Catalog No. 89110653 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-653 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-653 Supplier Abnova Corporation Supplier No. MAB1832

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against MAPT.

Specifications

Antigen MAPT
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TAU-5
Conjugate Unconjugated
Description microtubule-associated protein tau
Formulation In PBS, pH 7.3 (0.09% sodium azide)
Gene MAPT
Gene Alias MGC134287, tau
Gene Symbols MAPT
Host Species Mouse
Immunogen Bovine MAPT.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281296
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.