Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mast cell tryptase, Mouse, Clone: AA1, Abnova™
Mouse monoclonal antibody raised against native Mast cell tryptase.
Supplier: Abnova Corporation MAB5339
Specifications
Mast cell tryptase | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10000) The optimal working dilution should be determined by the end user. | |
Mouse | |
200 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
AA1 | |
Mouse monoclonal antibody raised against native Mast cell tryptase. | |
In PBS (0.09% sodium azide) | |
Native purified Mast cell tryptase from human lung tissue. | |
RUO | |
This antibody recognizes human mast cell tryptase, both alpha and beta isoforms. It is an excellent marker for mast cells, and does not bind to any other cell type in immunohistology. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction