Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAT2A, Mouse, Clone: AT3A2, Abnova™

Catalog No. 89111264 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-264 Supplier Abnova Corporation Supplier No. MAB3213
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant MAT2A.

Specifications

Antigen MAT2A
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone AT3A2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant MAT2A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene MAT2A
Gene Alias MATA2/MATII/SAMS2
Gene Symbols MAT2A
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human MAT2A.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4144
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.