Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MATK, Mouse, Clone: 9D7, Abnova™

Catalog No. 89109004 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-004 Supplier Abnova Corporation Supplier No. MAB10803
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant MATK.

The protein encoded by this gene has amino acid sequence similarity to Csk tyrosine kinase and has the structural features of the CSK subfamily: SRC homology SH2 and SH3 domains, a catalytic domain, a unique N terminus, lack of myristylation signals, lack of a negative regulatory phosphorylation site, and lack of an autophosphorylation site. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer. Three alternatively spliced transcript variants that encode different isoforms have been described for this gene. [provided by RefSeq

Specifications

Antigen MATK
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 9D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant MATK.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.03% sodium azide)
Gene MATK
Gene Alias CHK/CTK/DKFZp434N1212/HHYLTK/HYL/HYLTK/Lsk/MGC1708/MGC2101
Gene Symbols MATK
Host Species Mouse
Immunogen Recombinant protein corresponding to human MATK.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4145
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.