Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MB, Mouse, Clone: 34h5, Abnova™

Catalog No. 89113626 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-626 Supplier Abnova Corporation Supplier No. MAB8867
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant MB.

This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a haemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq

Specifications

Antigen MB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 34h5
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant MB.
Dilution The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene MB
Gene Alias MGC13548/PVALB
Gene Symbols MB
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human MB.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4151
Test Specificity This antibody recognizes recombinant protein MGB.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.