Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP, Mouse, Clone: BDI221, Abnova™

Catalog No. 89111751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-751 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-751 Supplier Abnova Corporation Supplier No. MAB4269
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against MBP.

The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called Golli-MBP) that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes. [provided by RefSeq]

Specifications

Antigen MBP
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone BDI221
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against MBP.
Dilution ELISA (10-20 ug/mL) Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) Immunohistochemistry (Frozen sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene MBP
Gene Alias MGC99675
Gene Symbols MBP
Host Species Mouse
Immunogen Purified human MBP.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4155
Test Specificity Reacts with residues 130-136 in human myelin basic protein (21.5KDa and 18.5KDa molecular forms). Recognizes primate, rabbit, sheep, goat, rat and mouse myelin basic protein.
Target Species Goat, Human, Mouse, Primate, Rabbit, Rat, Sheep
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.