Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP (Maltose Binding Protein), Mouse, Clone: 10C12, Abnova™

Catalog No. 89134856 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-856 Supplier Abnova Corporation Supplier No. MAB1133
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant MBP (Maltose Binding Protein).

Specifications

Antigen MBP (Maltose Binding Protein)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant MBP (Maltose Binding Protein).
Dilution Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to full length MBP (Maltose Binding Protein).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.