Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MC3R, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-892 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-892 Supplier Abnova Corporation Supplier No. H00004159A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MC3R.

This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans. [provided by RefSeq

Sequence: MSIQKTYLEGDFVFPVSSSSFLRTLLEPQLGSALLTAMNASCCLPSVQPTLPNGSEHLQAPFFSNQSSSAFCEQ

Specifications

Antigen MC3R
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MC3R.
Formulation 50% glycerol
Gene MC3R
Gene Accession No. NM_019888
Gene Alias BMIQ9/MC3/MC3-R
Gene Symbols MC3R
Host Species Mouse
Immunogen MC3R (NP_063941, 1 a.a. ∽ 74 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4159
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.