Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM, Mouse, Clone: 3H6, Abnova™

Catalog No. 89104893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-893 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-893 Supplier Abnova Corporation Supplier No. H00004162M19
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MCAM.

Sequence: VPGEAEQPAPELVEVEVGSTALLKCGLSQSQGNLSHVDWFSVHKEKRTLIFRVRQGQGQSEPGEYEQRLSLQDRGATLALTQVTPQDERIFLCQGKRPRSQEYRIQLRVYKAPEEPNIQVNPLGIPVNSKEPEEVATCVGRNGYPIPQVIWYKNGRPLKEEKNRVHIQSSQTVESSGLYTLQSILKAQLVKEDKDAQFYCELNYRLPSGNHMKESREVTVPVFYPTEKVWLEVEPVGMLKEGDRVEIRCLADGNPPPHFSISKQNPSTREAEEETTNDNGVLVLEPARKEHSGRYECQGLDLDTMISLLSEPQELLVNYVSDVRVSPAAPERQEGSSLTLTCEAESSQDLEFQWLREETGQVLERGPVLQLHDLKREAGGGYRCVASVPSIPGLNRTQLVNVAIFGPPWMAFKERKVWVKENMVLNLSCEASGHPRPTISWNVNGTASEQDQDPQRVLSTLNVLVTPELLETGVECTASNDLGKNTSILFLELVNLTTLTPDSNTTTGLSTSTASPHTRANSTSTERKLPEPESRGVVIVAVIVCILVLAVLGAVLYFLYKKGKLPCRRSGKQEITLPPSRKSELVVEVKSDKLPEEMGLLQGSSGDKRAPGDQGEKYIDLRH

Specifications

Antigen MCAM
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3H6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant MCAM.
Formulation PBS with no preservative; pH 7.4
Gene MCAM
Gene Accession No. NM_006500.2
Gene Alias CD146/MUC18
Gene Symbols MCAM
Host Species Mouse
Immunogen MCAM (NP_006491.2,24 a.a. ∽ 646 a.a) recombinant protein with his-flag-strepII tag.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4162
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.