Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAM, Mouse, Clone: C264, Abnova™
Mouse monoclonal antibody raised against native MCAM.
Supplier: Abnova Corporation MAB3278
Specifications
MCAM | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
MCAM | |
Mouse | |
500 μg | |
Primary | |
Chicken MCAM, Mr 98 KDa (protein core ∼84 KDa). | |
Store at 4°C. | |
IgG2b |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) | |
C264 | |
Mouse monoclonal antibody raised against native MCAM. | |
In 100mM BBS, pH 8.2 | |
MCAM | |
Native purified chicken MCAM. | |
RUO | |
448832 | |
Chicken | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction